Learn More
Miele Professional™ Water purification
ConductivityMeter for measuring the conductivity level and the display for changing the cartridge
Brand: Miele Professional™ 10008360
Additional Details : Weight : 2.48000kg
Description
- Suitable for full demineralisation cartridges
- Optical and acoustic display for cartridge change
- 10 switch points in measuring range 0 - 199.9 µS/cm
- One-line display, connection to external LED possible
- Including wall bracket and power unit
Specifications
Plastic | |
ConductivityMeter for measuring the conductivity level and the display for changing the cartridge | |
Water Pre-treatment | |
Plastic | |
110 mm | |
2.31 kg |
Black | |
Washer-disinfectors | |
Yes | |
125 mm | |
52 mm |