Learn More
Macherey-Nagel™ Visocolor™ ECO Iron 2 Colorimetric Test Kit
Brand: Macherey-Nagel™ 931026
Additional Details : Weight : 5.00000kg
Specifications
Natural waters as well as waste waters often contain some iron | |
0 to 1mg/L Fe | |
ECO Iron 2 Colorimetric Test Kit |
1 Each | |
100 Determinations |