Learn More
Macherey-Nagel™ Visocolor™ ECO Chromium (VI) Colorimetric Test Kit
Brand:  Macherey-Nagel™ 931020
Additional Details : Weight : 0.07250kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Product Code. 10645971
  • 725.00 NOK / 1 test
Estimated Shipment: 08-05-2024
Specifications
Chromium compounds are present in trivalent form [chromium (III) ions] and in hexavalent form (chromate and dichromate ions) in industrial waste water
0.02 to 0.5mg / l Cr (VI)
ECO Chromium (VI) Colorimetric Test Kit
1 Each
140 Provisions
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • VISOCOLOR ECO Chromium (VI)
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : o-phosphoric acid, dimethyl sulfoxide