Learn More
Macherey-Nagel™ Visocolor™ Alpha Nitrate Test Kit
Provides a fast, reliable and safe colorimetric determination of nitrate in samples. Macherey Nagel™ Visocolor™ Alpha Nitrate Test Kits are environmentally friendly, without hazardous reactants.
Brand:  Macherey-Nagel™ 935065
Product Code. 10073111
  • 507.00 NOK / Each
Explore more special offers
Description
Description
Provides a fast, reliable and safe colorimetric determination of nitrate in samples. Macherey Nagel™ Visocolor™ Alpha Nitrate Test Kits are environmentally friendly, without hazardous reactants.
  • With pictographic instructions
  • Reagent bottles with clear dosing instructions
  • Convenient handling, as easy as dip sticks
  • For the determination of nitrate in surface and drinking water and industrial waste waters
Specifications
Specifications
For the determination of nitrate in surface and drinking water and in industrial waste waters which do not contain high concentrations of interfering ions
2 to 50mg/L NO3-
Alpha Nitrate Colorimetric Test Kit
1 Each
100 Determinations
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • VISOCOLOR alpha Nitrate
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contains : 1-naphthylamine, acetic acid / sulfanilic acid, zinc powder(stabilizied)