Learn More
Vinyloxytrimethylsilane, 97%
Product Code. 11387059
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11387059
Brand: Thermo Scientific Alfa Aesar H26876.03
Request Bulk or Custom Format
CAS: 6213-94-1 | C5H12OSi | 116.235 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6213-94-1
Molecular Formula C5H12OSi
Molecular Weight (g/mol) 116.235
MDL Number MFCD00054764
InChI Key HFTNNOZFRQLFQB-UHFFFAOYSA-N
Synonym trimethyl vinyloxy silane, vinyloxytrimethylsilane, silane, ethenyloxy trimethyl, tmso-ethene, vinyloxy-trimethylsilane, trimethylsiloxy ethylene, snqhhgciiezjp@, acmc-209mzx, ethenoxy trimethyl silane, trimethylsilyl vinyl ether
PubChem CID 80344
IUPAC Name ethenoxy(trimethyl)silane
SMILES C[Si](C)(C)OC=C
Specifications
Boiling Point 74°C
Quantity 1 g
UN Number UN1993
Flash Point −19°C (−2°F)
Formula Weight 116.24
Percent Purity 97%
Refractive Index 1.389
Sensitivity Moisture sensitive
Density 0.779
Chemical Name or Material Vinyloxytrimethylsilane