missing translation for 'onlineSavingsMsg'
Learn More

Vinculin Antibody (VCL/7091R) [Janelia Fluor™ 669], Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals (Bio-Techne) NBP3-14197JF669

5261.50 NOK valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.


Product Code. 30171550

  • 6060.00 NOK / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Vinculin Monoclonal antibody specifically detects Vinculin in Human,Mouse samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Vinculin
Monoclonal
Janelia Fluor 669
CMD1W, CMH15, Metavinculin, MVCL, vinculin
Recombinant fragment (around aa 174-322) of human Vinculin (VCL) protein (exact sequence is proprietary) (Uniprot: P18206)
0.1 mL
Cell Biology, Cellular Markers, Phospho Specific
7414
Store at 4°C in the dark.
IgG
Western Blot, Immunohistochemistry (Paraffin)
VCL/7091R
50mM Sodium Borate
Rabbit
Protein A or G purified
RUO
Primary
Human, Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.