Learn More
Thermo Scientific™ VersiPlate Frame, 96-well, skirted, white
Flexible 96-well plates of low profile strip tubes for use in PCR and qPCR applications.
Brand: Thermo Scientific™ AB1805
Additional Details : Weight : 0.75000kg
Description
The Thermo Scientific VersiPlates are unique, with each strip of eight tubes being linked to each other forming the familiar 12 x 8 or 96-well ANSI format. The small tear points between strips enable a single strip or as many as required for the individual experiment to be separated.
Highlights- Easier sample handling as multiple strips of samples can be handled simultaneously
- Reduced consumables waste as only the required numbers of strips need to be removed with the remainder being stored for the next experiment
- Can be used in instruments with or without a VersiFrame
- The reusable polycarbonate frame can serve as a useful rack for a VersiPlate or any number of low profile strip tubes and facilitates sample loading into thermal cyclers
- The low profile format of the VersiPlate has a lower dead volume which reduces sample evaporation
- Available in white which improves signal reflection, reduces variation and increases the sensitivity of qPCR assays
- Can be used with all Thermo Scientific VersiCap™ Mats, cap strips, adhesive and heat seals
- PCR
- qPCR/Real-Time PCR
Specifications
No Barcode | |
VersiPlate | |
Frame | |
Polycarbonate | |
96-Well Plates |
25 Plate | |
White | |
Real-Time PCR Systems or Thermal Cyclers | |
96 | |
Low profile |