Learn More
VEGF Antibody (varisacumab) [Janelia Fluor™ 669], Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-28776JF669
3217.25 NOK valid until 2025-12-16
Use promo code "24090" to get your promotional price.
Product Code. 30171709
  • 3790.00 NOK / 0.10mL
Description
VEGF Monoclonal antibody specifically detects VEGF in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VEGF
Monoclonal
Janelia Fluor 669
MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor
VEGF
0.1 mL
Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cellular Markers, Cytokines and Growth Factors, Developmental Biology, Glycobiology, HIF Target Genes, Hypoxia, Metastasis, Neuroscience, Neurotransmission, Sensory Systems, Signal Transduction, Vision
7422
Store at 4°C in the dark.
IgG1
Flow Cytometry, ELISA, Functional Assay
varisacumab
50mM Sodium Borate
Human
Protein A purified