Learn More
Fisherbrand™ Vacuum Aspiration Advantage System Accessory, Table Top Stand
Designed for use with Fisherbrand™ Vacuum Aspiration Advantage Systems.
Brand: Fisherbrand™
Additional Details : Weight : 0.15000kg
Description
- Table top stand
- Used with HandVac and accessories for Vacuum Aspiration Advantage Systems
Fluids can be removed easily and very precisely from slides, petri dishes, microplates, cell culture containers or similar vessels by using different pipettes or glass tips which can be easily connected to the HandVac pipettor attachment.
Specifications
Aspiration Advantage System | |
Table Top Stand |