Learn More
Ursodeoxycholic Acid-24-13C, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Ursodeoxycholic Acid 24-13 C |
| CAS | 63296-46-8 |
| Molecular Formula | C23(13 C)H40O4 |
| Quantity | 0.05 g |
| Synonym | Cholan-24-oic-24-13 C acid, 3,7-dihydroxy-, (3α,5β,7β)- (9 CI), Ursodeoxycholic acid 24-13 C, Ursodeoxycholic-24-13 C acid, 3α,7β-Dihydroxy-5β-cholan-24-oic acid-24-13 C |
| InChI Formula | InChI=1 S/C24H40O4/c1-14(4-7-21(27)28)17-5-6-18-22-19(9-11-24(17,18)3)23(2)10-8-16(25)12-15(23)13-20(22)26/h14-20,22,25-26 H,4-13H2,1-3H3,(H,27,28)/t14-,15+,16-,17-,18+,19+,20+,22+,23+,24-/m1/s1/i21+1 |
| SMILES | C[C@@]12 [C@](CC[C@]2([H])[C@H](C)CC[13 C](O)=O)([H])[C@@]3([H])[C@@](CC1)([H])[C@@]4([C@](C[C@@H](CC4)O)([H])C[C@@H]3 O)C |
| IUPAC Name | (R)-4-((3 R,5 S,7 S,8 R,9 S,10 S,13 R,14 S,17 R)-3,7-dihydroxy-10,13-dimethylhexadecahydro-1 H-cyclopenta[a]phenanthren-17-yl)pentanoic-1-13 C acid |
| Molecular Weight (g/mol) | 393.57 |
| Formula Weight | 393.3 g/mol |
| Show More |