missing translation for 'onlineSavingsMsg'
Learn More

Uranium solution for ICP/MS 10mg/L HNO3 matrix, Fisher Chemical™

Catalog Number p-7115370
Click to view available options
Quantity:
50 mL
100 mL
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
U
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
U

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Concentration 10mg/L
Packaging Bottle
Quantity 50 mL
Solvent or Matrix HNO3
Chemical Name or Material Uranium
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'