Learn More
Miele Professional™ Insert A 11/1
Insert A 11/1 for the gentle cleaning of delicate items by interrupting the spray jet
Brand: Miele Professional™ 6629660
Additional Details : Weight : 0.50000kg
Description
- With perforations 7 x 7 mm
- For use in upper and lower basket
Specifications
Underlay Frame | |
450 x 450mm Inserts. | |
1 Each |