missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBP1 Mouse anti-Human, Unconjugated, Clone: 1B10H3, Proteintech
Mouse Monoclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | UBP1 |
---|---|
Clone | 1B10H3 |
Concentration | 1000 μg/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16889875
|
Proteintech
67318-1-IG-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16879875
|
Proteintech
67318-1-IG-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Descrizione
The UBP1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human UBP1. This antibody recognizes human antigen. The UBP1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifica
UBP1 | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
7342 | |
Primary |
1B10H3 | |
Western Blot, ELISA | |
Unconjugated | |
Human | |
IgG1 |