Learn More
Dostmann Electronic™ Type K Thermo Couple Immersion
Product Code. 10015961
Change view
Click to view available options
Temperature Range (Metric):
-200°C to +1100°C
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Temperature Range (Metric) unitSize
10015961 -200°C to +1100°C Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10015961 Supplier Dostmann Electronic™ Supplier No. 60101006
  • ONLINE EXCLUSIVE PRICE
    1123.50 NOK
    795.52 NOK / Pack of 1
    Save 327.98 NOK
    29% Off
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
Order Info
LengthMetric
Specifications
Type Thermocouple Immersion
Material PVC
Sensor Types Class 1
Temperature Range (Metric) -200°C to +1100°C
Length (Metric) 300 mm