Learn More
Turofexorate isopropyl, MedChemExpress
Product Code. 16487896
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16487896
Brand: Medchem Express HY50911
MedChemExpress Turofexorate isopropyl (FXR-450) is a potent, selective, and orally bioavailable FXR agonist with EC50 of 4 nM.
Turofexorate isopropyl (FXR-450) is a potent, selective, and orally bioavailable FXR agonist with EC50 of 4 nM.
Specifications
Chemical Name or Material Turofexorate isopropyl
CAS 629664-81-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C25H24F2N2O3
Quantity 100 mg
Synonym FXR-450 XL335 WAY-362450
Solubility Information DMSO : 25 mg/mL (57.02 mM; Need ultrasonic)
SMILES O=C(C1=CN(C(C2=CC=C(F)C(F)=C2)=O)CC(C)(C)C3=C1NC4=C3C=CC=C4)OC(C)C
Molecular Weight (g/mol) 438.47
Formula Weight 438.47
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.