Learn More
Masterflex™ Tubing Adapter, PVDF Female Luer Lock
Standard connection type for syringes, plus ideal for many applications with small tubing.
Brand: Masterflex™ 31201-50
Additional Details : Weight : 0.01000kg
Description
These Female luer lock x 3/4 in. mini sanitary adapters are available in either polypropylene or PVDF.Specifications
Vacuum Tubing | |
Sanitary x Luer | |
PVDF |
Sanitary Tubing Adapter, PVDF; 3/4 in. Mini to Female Luer Lock | |
Sanitary Clamp Size (Port 1): 3/4 in. |