Learn More
Masterflex™ Tubing adapter NPT (M) PVDF
Ideal for general-purpose and single-use applications, to connect tubing with rest of system
Brand:  Masterflex™ 31201-00
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12458670
Quantity Price
1 1452.00 NOK / Each
10 1355.44 NOK / Each
Estimated Shipment: 27-09-2021
Add to basket
Description
Description
  • Determine which connector type is needed for your application by contacting free technical application assistance
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Always in stock—most materials and configurations available to ship same day
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Meet USP Class VI for pharmaceutical use and FDA, USDA, and 3A standards for food contact
  • All fittings are compatible with Tri-Clamp™ connections
  • Excellent chemical resistance; low leachable characteristics
Specifications
Specifications
Tubing Adapter; , 3/4 in. mini to 1/8 in. NPT (M)
Masterflex L/S and I/P sanitary tubing connection
1/8 in.
PVDF
Vacuum Tubing
Sanitary x NPT(M)
Connection Type: 3/4 in. mini to 1/8 in. NPT(M)