missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TTC31 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21693-1-AP-20UL
Additional Details : Weight : 0.00010kg
Specifications
TTC31 | |
Polyclonal | |
Unconjugated | |
TTC31 | |
TPR repeat protein 31, TTC31 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
64427 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q49AM3 | |
TTC31 | |
TTC31 Fusion Protein Ag16281 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |