missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Troponin T Type 2 (cardiac) Antibody (1F11cc) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB120-10214JF585
This item is not returnable.
View return policy
Description
Troponin T Type 2 (cardiac) Monoclonal antibody specifically detects Troponin T Type 2 (cardiac) in Bovine,Canine,Feline,Fish,Goat,Human,Mouse,Porcine,Rabbit,Rat samples. It is validated for Western Blot,Sandwich ELISA
Specifications
| Troponin T Type 2 (cardiac) | |
| Monoclonal | |
| Janelia Fluor 585 | |
| cardiac, CMD1D, CMPD2, dilated 1D (autosomal dominant), hypertrophic 2, MGC3889, RCM3, TnTc, troponin T type 2 (cardiac), troponin T, cardiac muscle | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with free human cTnT. | |
| 0.2 mL | |
| Cell Biology, Cellular Markers | |
| 7139 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, Sandwich ELISA | |
| 1F11cc | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Bovine, Canine, Feline, Fish, Goat, Human, Mouse, Pig, Rabbit, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction