Learn More
Trimethyl borate, 99%, AcroSeal™, Thermo Scientific Chemicals
Catalog Number p-4089430
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
121-43-7
C3H9O3B
103.91
MFCD00008346
WRECIMRULFAWHA-UHFFFAOYSA-N
trimethoxyborane, methyl borate, trimethoxyboron, boric acid, trimethyl ester, trimethylborate, boric acid trimethyl ester, trimethoxyborine, borester o, boric acid h3bo3 , trimethyl ester, b och3 3
8470
CHEBI:38913
trimethyl borate
B(OC)(OC)OC
121-43-7
C3H9O3B
103.91
MFCD00008346
WRECIMRULFAWHA-UHFFFAOYSA-N
trimethoxyborane, methyl borate, trimethoxyboron, boric acid, trimethyl ester, trimethylborate, boric acid trimethyl ester, trimethoxyborine, borester o, boric acid h3bo3 , trimethyl ester, b och3 3
8470
CHEBI:38913
trimethyl borate
B(OC)(OC)OC
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point -34.0°C
Density 0.9150g/mL
Boiling Point 68.0°C to 69.0°C
Flash Point −8°C
Assay Percent Range 99%
Packaging AcroSeal™ Glass Bottle
Linear Formula (CH3O)3B
Refractive Index 1.3562 to 1.3582
Quantity 100 mL
Beilstein 01, 287
Fieser 01,673; 13,218
Merck Index 15, 9882
Specific Gravity 0.915
Solubility Information Solubility in water: reacts. Other solubilities: miscible with thf,diethyl ether,hexane,methanol,,isopropylamine,and other organic liquids
Viscosity 0.36 mPa.s (25°C)
Formula Weight 103.91
Percent Purity 99%
Chemical Name or Material Trimethyl borate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trimethyl borate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
Supplemental information
  • MIXTURE LIST-Contains : Trimethyl borate, methyl alcohol