missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tricyclohexylphosphonium tetrafluoroborate, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Tricyclohexylphosphonium tetrafluoroborate is used with ruthenium (1,5-cyclooctadiene)ruthenium dimer to catalyze the dehydrogenative coupling of alcohols and amines to form amide bonds. It is also used in suzuki reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tricyclohexylphosphonium tetrafluoroborate is used with ruthenium (1,5-cyclooctadiene)ruthenium dimer to catalyze the dehydrogenative coupling of alcohols and amines to form amide bonds. It is also used in suzuki reaction.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Tricyclohexylphosphonium tetrafluoroborate is used with ruthenium (1,5-cyclooctadiene)ruthenium dimer to catalyze the dehydrogenative coupling of alcohols and amines to form amide bonds. It is also used in suzuki reaction.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | 166°C to 170°C |
Linear Formula | (C6H11)3PHBF4 |
Quantity | 1 g |
Solubility Information | Soluble in water. |
Formula Weight | 368.25 |
Percent Purity | 99% |
Chemical Name or Material | Tricyclohexylphosphonium tetrafluoroborate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'