missing translation for 'onlineSavingsMsg'
Learn More

Tricarbonylnitrosylcobalt, Thermo Scientific Chemicals

Catalog Number p-7049822
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
14096-82-3
MFCD00016014
cobalt tricarbonyl nitrosyl, tricarbonyl-nitrosyl-cobalt
This item is not returnable. View return policy
14096-82-3
MFCD00016014
cobalt tricarbonyl nitrosyl, tricarbonyl-nitrosyl-cobalt

Tricarbonylnitrosylcobalt is used as the Co metalorganic precursor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -1°C
Color Red
Density 1.47 g/mL
Boiling Point 48.6°C (55°C (decomposition))
Odor Unpleasant
Linear Formula Co(NO)(CO)3
Quantity 1 g
UN Number UN1992
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 172.97
Physical Form Liquid
Chemical Name or Material Tricarbonylnitrosylcobalt
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'