Learn More
Uvex Trend Black/Gray/Red PU/PU 69097 Low Shoe - Width 10
Uvex 2 Trend safety footwear for medium-risk applications (S1-S3); heel cap offers increased stability without limiting movement
1105.00 NOK
Specifications
Color | Black, Gray, Red |
---|---|
Material | Microvelour |
Sole Material | Dual-density polyurethane (PU/PU) |
Toe Material | Steel |
Disposable | No |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17203331
|
Uvex
6909738 |
38 |
1105.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17213331
|
Uvex
6909739 |
39 |
1105.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17223331
|
Uvex
6909740 |
40 |
1105.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17233331
|
Uvex
6909741 |
41 |
1105.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17243331
|
Uvex
6909742 |
42 |
1105.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17253331
|
Uvex
6909743 |
43 |
1105.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17263331
|
Uvex
6909744 |
44 |
1105.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17273331
|
Uvex
6909745 |
45 |
1105.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17283331
|
Uvex
6909746 |
46 |
1105.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17293331
|
Uvex
6909747 |
47 |
1105.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17203341
|
Uvex
6909748 |
48 |
1105.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17213341
|
Uvex
6909749 |
49 |
1105.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17223341
|
Uvex
6909750 |
50 |
1105.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17233341
|
Uvex
6909751 |
51 |
1105.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
17243341
|
Uvex
6909752 |
52 |
1105.00 NOK
1 pair |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
Description
Uvex 2 Trend Shoe 69097 S2 SRC
- Sporty, S2 safety shoe with wide fit
- Breathable, water repellent perforated microvelour
- Made from synthetic materials, suitable for people allergic to chrome
- All sole materials free of silicones, plasticizers and other paint wetting impairment substances
- Meets ESD specification with volume resistance under 35 megohms
- Reliable protection for wearers
- Steel toe cap and stabilizing Uvex anti-twist heel cap to prevent ankle twisting
- Ergonomically designed outsole made from dual-density polyurethane with excellent slip resistance
- Reflective details for enhanced visibility
- Uvex Climazone technology lining, upper and removable comfortable climatic insole with a moisture transport system
- Helps maintain foot temperature for optimum wearer comfort
- Soft padding on collar and tongue
- Removable anti-static comfortable insole with moisture transport system and additional shock absorption around heel and forefoot
- Article No. 69097
- Standard EN ISO 20345:2011 S2 SRC
- Sizes: 38 to 52
Specifications
Black, Gray, Red | |
Dual-density polyurethane (PU/PU) | |
No | |
Unisex | |
Uvex 2 | |
No | |
SRC |
Microvelour | |
Steel | |
EN ISO 20345:2011 | |
Narrow width | |
Yes | |
S2 |