Learn More
Uvex Trend Black/Gray PU/PU 69461 Sandal - Width 10
Uvex 2 Trend safety footwear for medium-risk applications (S1-S3); heel cap offers increased stability without limiting movement
1105.00 NOK
Specifications
Color Black, Gray
Material Microvelour
Sole Material Dual-density polyurethane (PU/PU)
Toe Material Steel
Disposable No
View More Specs
Products 15
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
17233981
View Documents Promotion Details
Uvex
6946138
38
1105.00 NOK
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 22-05-2024
to see stock.
Add to basket
17243981
View Documents Promotion Details
Uvex
6946139
39
1105.00 NOK
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 22-05-2024
to see stock.
Add to basket
17253981
View Documents Promotion Details
Uvex
6946140
40
1105.00 NOK
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 22-05-2024
to see stock.
Add to basket
17263981
View Documents Promotion Details
Uvex
6946141
41
1105.00 NOK
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 22-05-2024
to see stock.
Add to basket
17273981
View Documents Promotion Details
Uvex
6946142
42
1105.00 NOK
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 22-05-2024
to see stock.
Add to basket
17283981
View Documents Promotion Details
Uvex
6946143
43
1105.00 NOK
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 22-05-2024
to see stock.
Add to basket
17293981
View Documents Promotion Details
Uvex
6946144
44
1105.00 NOK
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 22-05-2024
to see stock.
Add to basket
17203991
View Documents Promotion Details
Uvex
6946145
45
1105.00 NOK
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 22-05-2024
to see stock.
Add to basket
17213991
View Documents Promotion Details
Uvex
6946146
46
1105.00 NOK
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 22-05-2024
to see stock.
Add to basket
17223991
View Documents Promotion Details
Uvex
6946147
47
1105.00 NOK
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 22-05-2024
to see stock.
Add to basket
17233991
View Documents Promotion Details
Uvex
6946148
48
1105.00 NOK
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 22-05-2024
to see stock.
Add to basket
17243991
View Documents Promotion Details
Uvex
6946149
49
1105.00 NOK
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 22-05-2024
to see stock.
Add to basket
17253991
View Documents Promotion Details
Uvex
6946150
50
1105.00 NOK
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 22-05-2024
to see stock.
Add to basket
17263991
View Documents Promotion Details
Uvex
6946151
51
1105.00 NOK
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 22-05-2024
to see stock.
Add to basket
17273991
View Documents Promotion Details
Uvex
6946152
52
1105.00 NOK
1 pair
View alternative products
Save Up to 
»
Estimated Shipment: 22-05-2024
Description
Uvex 2 Trend Sandal 69461 S1 P SRC
  • Sporty S1 P safety sandal with wide fit
  • Breathable microvelour
  • Made from synthetic materials, suitable for people allergic to chrome
  • All sole materials free of silicones, plasticizers and other paint wetting impairment substances
  • Double, individually adjustable
  • Counter-traction hook and loop straps
  • Meets ESD specification with volume resistance under 35 megohms
  • Reliable protection for wearers
  • Penetration-resistant steel midsole, steel toecap and stabilizing Uvex anti-twist heel cap to prevent ankle twisting
  • Ergonomically designed outsole comprising dual-density polyurethane with excellent slip resistance
  • Reflective details for enhanced visibility
  • Uvex Climazone technology lining, upper and removable comfortable insole
    • Moisture transport system helps maintain foot temperature
  • Soft padding on collar and dust tongue
  • Removable anti-static comfortable insole with a moisture transport system and additional shock absorption around heel and forefoot
  • Article No. 69461
  • Standard EN ISO 20345:2011 S1 P SRC
  • Sizes: 38 to 52
Specifications
Specifications
Black, Gray
Dual-density polyurethane (PU/PU)
No
Unisex
Uvex 2
No
SRC
Microvelour
Steel
EN ISO 20345:2011
Narrow width
Yes
S1