missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ TREM2 Recombinant Protein

Human TREM2 full-length ORF recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00054209-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16140493

  • 6245.00 NOK / 25µg
Estimated Shipment: 27-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is a membrane protein that forms a receptor signaling complex with TYROBP. The encoded protein may be involved in chronic inflammation by triggering the production of constitutive inflammatory cytokines. Defects in this gene are a cause of polycystic lipomembranous osteodysplasia with sclerosing leukoencephalopathy (PLOSL)

  • Theoretical MW (kDa): 50.16
  • Preparation method: In vitro wheat germ expression system
  • Purification: Glutathione Sepharose 4 fast flow
  • Storage buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer

Sequence: MEPLRLLILLFVTELSGAHNTTVFQGVAGQSLQVSCPYDSMKHWGRRKAWCRQLGEKGPCQRVVSTHNLWLLSFLRRWNGSTAITDDTLGGTLTITLRNLQPHDAGLYQCQSLHGSEADTLRKVLVEVLADPLDHRDAGDLWFPGESESFEDAHVEHSISRPSQGSHLPSCLSKEPLGRRNPLPTHFHPSPPGLHLSHQDSSSQRPLGCSLAWTEARDTSTQ

Best when used within three months from the date of receipt.

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH32362
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
50.16
8
Glutathione Sepharose 4 Fast Flow
25 ug
MEPLRLLILLFVTELSGAHNTTVFQGVAGQSLQVSCPYDSMKHWGRRKAWCRQLGEKGPCQRVVSTHNLWLLSFLRRWNGSTAITDDTLGGTLTITLRNLQPHDAGLYQCQSLHGSEADTLRKVLVEVLADPLDHRDAGDLWFPGESESFEDAHVEHSISRPSQGSHLPSCLSKEPLGRRNPLPTHFHPSPPGLHLSHQDSSSQRPLGCSLAWTEARDTSTQ
TREM-2/Trem2a/Trem2b/Trem2c
TREM2
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
54209
TREM2 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TREM2
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers