missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TREM2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1415.13 NOK - 3828.53 NOK
Specifications
Antigen | TREM2 |
---|---|
Concentration | 0.37 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16801724
|
Proteintech
13483-1-AP-20UL |
20 μL |
1415.13 NOK
20µL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16891714
|
Proteintech
13483-1-AP-150UL |
150 μL |
3828.53 NOK
150µL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
TREM2 is a membrane protein that forms a receptor signaling complex with TYROBP. The encoded protein may be involved in chronic inflammation by triggering the production of constitutive inflammatory cytokines. Defects in this gene are a cause of polycystic lipomembranous osteodysplasia with sclerosing leukoencephalopathy (PLOSL).Specifications
TREM2 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q99NH8, Q9NZC2 | |
54209, 83433 | |
TREM2 Fusion Protein Ag4299 | |
Primary | |
-20°C | |
TREM2 |
0.37 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
TREM 2, TREM2, Trem2a, Trem2b, Trem2c | |
TREM2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |