Learn More
Tenak™ Single Tray Rack for Upright Freezers, Wide
Trays designed for applications where the samples are either not stored in boxes or has odd sizes.
Brand:  Tenak™ TE29228
Additional Details : Weight : 0.55000kg
 View more versions of this product
Product Code. 12486990
  • 1175.00 NOK / Each
Estimated Shipment: 10-12-2021
Add to basket
Description
Description
  • Single trays are a drawer including a lid with a handle on the front and it will fit into any freezer
  • Racks can be used with all freezer brands
  • Constructed of stainless steel
Specifications
Specifications
560 x 278 x 280 mm
Upright Freezers
Stainless Steel
278 mm