missing translation for 'onlineSavingsMsg'
Learn More

trans-4-n-Propyl-4'-(3,4,5-trifluorophenyl)bicyclohexyl, 97%, Thermo Scientific™

Catalog Number p-7037129
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
131819-23-3
C21H29F3
338.458
MFCD09838998
FEWMLRARKGRCCE-UHFFFAOYSA-N
trans,trans-4'-propyl-4-3,4,5-trifluorophenyl bicyclohexyl, trans-4-3,4,5-trifluorophenyl-trans-4'-propylbicyclohexane, trans-4-n-propyl-4'-3,4,5-trifluorophenyl bicyclohexyl, benzene, 1,2,3-trifluoro-5-trans,trans-4'-propyl 1,1'-bicyclohexyl-4-yl, 4-3,4,5-trifluorophenyl-4'-propyl-1,1'-bicyclohexane, trans,trans-1-4'-propylbicyclohexyl-3,4,5-trifluorobenzene, 1,2,3-trifluoro-5-trans,trans-4-propyl 1,1-bicyclohexyl-4-yl benzene, all-trans-4-propyl-4'-3,4,5-trifluorophenyl bi cyclohexane, 1s,1'r,4r,4'r-4-propyl-4'-3,4,5-trifluorophenyl-1,1'-bi cyclohexane, 1s,4r-4-propyl-4'-3,4,5-trifluorophenyl-1,1'-bi cyclohexane
11772113
1,2,3-trifluoro-5-[4-(4-propylcyclohexyl)cyclohexyl]benzene
CCCC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C(=C3)F)F)F
This item is not returnable. View return policy
131819-23-3
C21H29F3
338.458
MFCD09838998
FEWMLRARKGRCCE-UHFFFAOYSA-N
trans,trans-4'-propyl-4-3,4,5-trifluorophenyl bicyclohexyl, trans-4-3,4,5-trifluorophenyl-trans-4'-propylbicyclohexane, trans-4-n-propyl-4'-3,4,5-trifluorophenyl bicyclohexyl, benzene, 1,2,3-trifluoro-5-trans,trans-4'-propyl 1,1'-bicyclohexyl-4-yl, 4-3,4,5-trifluorophenyl-4'-propyl-1,1'-bicyclohexane, trans,trans-1-4'-propylbicyclohexyl-3,4,5-trifluorobenzene, 1,2,3-trifluoro-5-trans,trans-4-propyl 1,1-bicyclohexyl-4-yl benzene, all-trans-4-propyl-4'-3,4,5-trifluorophenyl bi cyclohexane, 1s,1'r,4r,4'r-4-propyl-4'-3,4,5-trifluorophenyl-1,1'-bi cyclohexane, 1s,4r-4-propyl-4'-3,4,5-trifluorophenyl-1,1'-bi cyclohexane
11772113
1,2,3-trifluoro-5-[4-(4-propylcyclohexyl)cyclohexyl]benzene
CCCC1CCC(CC1)C2CCC(CC2)C3=CC(=C(C(=C3)F)F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
trans-4-n-Propyl-4′-(3,4,5-trifluorophenyl)bicyclohexyl is used to make PDLC mixtures and Switchable Film . It is also mainly used as a liquid crystal monomer, liquid crystal material.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 63°C to 64°C
Quantity 1 g
Solubility Information It is soluble in water.
Formula Weight 338.46
Percent Purity 97%
Chemical Name or Material trans-4-n-Propyl-4'-(3,4,5-trifluorophenyl)bicyclohexyl
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'