missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-3-(Benzyloxymethyl)cyclobutanol, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
trans-3-(Benzyloxymethyl)cyclobutanol is used as a raw material intermediates for pharmaceutical and chemical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
trans-3-(Benzyloxymethyl)cyclobutanol is used as a raw material intermediates for pharmaceutical and chemical.
Solubility
Slightly soluble in water.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
trans-3-(Benzyloxymethyl)cyclobutanol is used as a raw material intermediates for pharmaceutical and chemical.
Solubility
Slightly soluble in water.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
Specifications
Quantity | 250 mg |
Solubility Information | Slightly soluble in water. |
Formula Weight | 192.25 |
Percent Purity | 97% |
Chemical Name or Material | trans-3-(Benzyloxymethyl)cyclobutanol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'