missing translation for 'onlineSavingsMsg'
Learn More

trans-(1R,2R)-(-)-N,N'-Dimethyl-1,2-cyclohexanediamine dihydrochloride, 98%, Thermo Scientific™

Catalog Number p-4813689
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
70708-33-7
C8H18N2·2ClH
215.17
UDYXZFBJGBDHHP-RHJRFJOKSA-N
1r,2r-n1,n2-dimethylcyclohexane-1,2-diamine dihydrochloride, trans-n,n'-dimethyl-1,2-diaminocyclohexane dihydrochloride, 1,2-cyclohexanediamine, n,n'-dimethyl-, dihydrochloride, 1r-trans, trans-1r,2r---n,n'-dimethyl-1,2-cyclohexanediamine dihydrochloride, trans-n1,n2-dimethylcyclohexane-1,2-diamine dihydrochloride, 1r,2r-1-n,2-n-dimethylcyclohexane-1,2-diamine dihydrochloride, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexane diamine dihydrochloride, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexanediamine 2hcl, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexanediamine dihydrochloride, 1r,2r-n,n'-dimethyl-1,2-cyclohexanediamine dihydrochloride
44890801
(1R,2R)-1-N,2-N-dimethylcyclohexane-1,2-diamine;dihydrochloride
CNC1CCCCC1NC.Cl.Cl
This item is not returnable. View return policy
70708-33-7
C8H18N2·2ClH
215.17
UDYXZFBJGBDHHP-RHJRFJOKSA-N
1r,2r-n1,n2-dimethylcyclohexane-1,2-diamine dihydrochloride, trans-n,n'-dimethyl-1,2-diaminocyclohexane dihydrochloride, 1,2-cyclohexanediamine, n,n'-dimethyl-, dihydrochloride, 1r-trans, trans-1r,2r---n,n'-dimethyl-1,2-cyclohexanediamine dihydrochloride, trans-n1,n2-dimethylcyclohexane-1,2-diamine dihydrochloride, 1r,2r-1-n,2-n-dimethylcyclohexane-1,2-diamine dihydrochloride, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexane diamine dihydrochloride, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexanediamine 2hcl, trans-1r,2r-n,n'-bismethyl-1,2-cyclohexanediamine dihydrochloride, 1r,2r-n,n'-dimethyl-1,2-cyclohexanediamine dihydrochloride
44890801
(1R,2R)-1-N,2-N-dimethylcyclohexane-1,2-diamine;dihydrochloride
CNC1CCCCC1NC.Cl.Cl
Assay Percent Range 97.5 to max. 102.5 % min. (Argentometry)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 215.17
Percent Purity 98%
Chemical Name or Material trans-(1R,2R)-(-)-N,N'-Dimethyl-1,2-cyclohexanediamine dihydrochloride
compliance-icons
Product Identifier
  • trans-(1R,2R)-(-)-N,N'-Dimethyl-1,2-cyclohexanediamine dihydrochloride, 98%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'