Learn More
TRA-1-81 (Podocalyxin) Monoclonal Antibody (TRA-1-81), APC, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
1336.00 NOK - 3135.00 NOK
Specifications
Clone | TRA-1-81 |
---|---|
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | APC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15588206
|
Affymetrix eBioscience
17-8883-42 |
100 Tests |
3135.00 NOK
100 tests |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15578206
|
Affymetrix eBioscience
17-8883-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: The TRA-1-81 antibody recognizes a protein expressed on undifferentiated human embryonic stem cells (ES), embyronal carcinoma cells (EC), and embryonic germ cells (EG). Like other stem cell specific markers, the epitope recognized by the TRA-1-81 antibody is lost upon cell differentiation. The TRA-1-81 epitope is resistant to neuraminidase digestion, unlike the epitope recognized by the related TRA-1-60 antibody. The TRA-1-81 antibody is known to specifically recognize a carbohydrate epitope on a keratan sulfated glycoprotein recently identified as podocalyxin, a member of the CD34-related family of sialomucins. Podocalyxin is a transmembrane glycoprotein originally identified on epithelial glomerular cells known as podocytes, and the protein has also been implicated in the development of aggressiveness in a variety of cancers, including breast and prostate cancer. Applications Reported: This TRA-1-81 antibody has been reported for use in flow cytometric analysis. Applications Tested: This TRA-1-81 antibody has been pre-titrated and tested by flow cytometric analysis of 2102Ep human embryonal carcinoma cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.
TRA-1-81 (Podocalyxin) Monoclonal antibody specifically detects TRA-1-81 (Podocalyxin) in Human samples. It is validated for Flow Cytometry.Specifications
TRA-1-81 | |
Flow Cytometry | |
APC | |
Mouse | |
Human | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
IgM | |
Affinity chromatography | |
Antibody |
For Research Use Only.