missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TNFRSF6B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | TNFRSF6B |
---|---|
Concentration | 0.47 mg/mL |
Applications | Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16803525
|
Proteintech
24999-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16893515
|
Proteintech
24999-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by certain cytokines including TNF (tumor necrosis factor) and Fas ligand of the TNF family through their death domain containing receptors, TNFR1 (tumor necrosis factor receptor 1) and Fas. Cell death signals are transduced by death domain (DD)-containing adapter molecules and members of the ICE (interleukin-1-beta converting enzyme)/CED-3 (C. elegans death receptor-3) protease family. A novel DD-containing molecule was recently cloned from mouse, human and monkey and designated Daxx (death associated protein 6). Daxx binds specifically to the Fas death domain and enhances Fas induced apoptosis and activates the Jun N-terminal kinase (JNK) pathway. Daxx is widely expressed in fetal and adult human and mouse tissues indicating its important function in Fas signaling pathways.Specifications
TNFRSF6B | |
Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human | |
O95407 | |
8771 | |
TNFRSF6B Fusion Protein Ag19781 | |
Primary | |
-20°C | |
TNFRSF6B |
0.47 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
DCR3, Decoy receptor 3, Decoy receptor for Fas ligand, M68, TNFRSF6B, TR6 | |
TNFRSF6B | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |