Learn More
Alfa Aesar™ Titanium Thinfoil, 0.0127mm (0.0005 in.) thick, 99.6% (metals basis)
Brand: Alfa Aesar™ 044243.6Z
Additional Details : CAS Number : 7440-32-6 Weight : 25.00000kg
Quantity | 90 x 500mm |
---|
Chemical Identifiers
7440-32-6 | |
47.867 | |
RTAQQCXQSZGOHL-UHFFFAOYSA-N | |
23963 | |
titanium |
Ti | |
MFCD00011264 | |
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1 | |
CHEBI:33341 | |
[Ti] |
Specifications
Titanium Thinfoil | |
90 x 500mm | |
7440-32-6 | |
Ti | |
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1 | |
[Ti] | |
Odorless | |
23963 | |
Thin Foil | |
Commercial Pure 4 |
≈0.515g/90x100mm | |
0.0127mm (0.0005 in.) thick, Light Tight | |
(metals basis) | |
MFCD00011264 | |
RTAQQCXQSZGOHL-UHFFFAOYSA-N | |
titanium | |
47.867 | |
CHEBI:33341 | |
99.6% |