Learn More
Titanium aluminide, -325 mesh, 99.5% (metals basis)
Product Code. 11399259
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11399259
Brand: Thermo Scientific Alfa Aesar 088393.18
Request Bulk or Custom Format
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
CAS: 12003-96-2 | AlTi3+3 | 170.583 g/mol
It finds use in several applications including automobiles and aircraft. Also used in air frames, investment casting, turbocharger rotors, and wire/fiber metal matrix composites
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 12003-96-2
Molecular Formula AlTi3+3
Molecular Weight (g/mol) 170.583
MDL Number MFCD00210656
InChI Key JRDVYNLVMWVSFK-UHFFFAOYSA-N
Synonym aluminum 3+ ion trititanium, aluminum, compd. with titanium 1:1
PubChem CID 44153015
IUPAC Name aluminum;titanium
SMILES [Al+3].[Ti].[Ti].[Ti]
Specifications
Quantity 50 g
UN Number UN3089
Solubility Information Insoluble in water.
Percent Purity 99.5%
Odor Odorless
Sensitivity Air Sensitive
Assay (metals basis)
Chemical Name or Material Titanium aluminide