missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Tintometer™ VARIO™ Iron Ferro Reagent Powder, 25mL
Formulated for use in Lovibond test kits, colorimeters and spectrophotometers. Newly redesigned foil packs are easy to handle, easy to open and pour
Brand: Lovibond™ 530570
Additional Details : Weight : 0.30000kg
Description
- For use in our test kits, colorimeters and spectrophotometers
- Pre-measured, unit dose reagents
- Easy to handle, easy to open and easy to pour
- Case of 100
- Fast and easy use of VARIO Powder Packs has made them extremely popular for water testing applications in many countries throughout the world
- Provides more experienced users with a real alternative to existing measurement systems
- Produced to the same high quality standards that have made Tintometer’s tablet reagents so successful for several decades
- Parameters from aluminum and chlorine through to sulphate are just some of the well-known tests that are included in the VARIO Powder Pack range
Specifications
Water Testing | |
Case of 100 | |
VARIO™ Iron Ferro Reagent Powder |
Test kits, colorimeters and spectrophotometers | |
25 mL |