Learn More
Lovibond™ Tintometer™ Refill Pack, Chlorine pH
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
Brand:  Lovibond™ 515884
Additional Details : Weight : 0.30000kg
Product Code. 15381885
  • 159.00 NOK / Each
Estimated Shipment: 24-05-2024
Description
  • Reagent Refill pack for Pooltester
  • Contains reagents for 30 tests for each parameter:
  • 30 x DPD No. 1 Rapid tablet reagents
  • 30 x Phenol Red Rapid tablet reagents
Specifications
Specifications
Photometry Tablet Reagents
Test kits, colorimeters and spectrophotometers
30 x DPD No. 1 Rapid tablet reagents; 30 x Phenol Red Rapid tablet reagents
Refill Pack, Chlorine pH