missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Tintometer™ Molybdate No. 2 HR Tablet
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
550.00 NOK - 1145.00 NOK
Specifications
For Use With (Application) | Photometry Tablet Reagents |
---|---|
For Use With (Equipment) | Test kits, colorimeters and spectrophotometers |
Range | 1 to 50mg/L |
Type | Molybdate No. 2 HR Tablet |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15311785
|
Lovibond™
513070BT |
100 Tablets |
550.00 NOK
Pack of 100 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
15321785
|
Lovibond™
513071BT |
250 Tablets |
1145.00 NOK
Pack of 250 |
Estimated Shipment: 26-06-2024
Log in to see stock. |
|||||
Description
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
- Range: 1 to 50mg/L
- Parameter: Molybdate LR, molybdate, HR molybdate
- Method: Thioglycolate
- For use in our test kits, colorimeters and spectrophotometers
- New aluminum foil blister packaging brings added convenience to the tradition of protection achieved in Lovibond’s long established tablet production technology
- With the new blister strip, the user just pushes the tablet through the protective foil, straight into the sample
- Each tablet is contained within an individually formed foil cup, lined with the latest aluminum composite material, and guaranteeing product performance
- Blister pack has been reduced in size to 91 x 34mm for convenient storage and shipping
- ‘BT’ is added to the end of the code to identify the new style of packaging
- Simple, time-saving and practical
- There are no safety risks if the tablets are used in line with the instructions supplied
- Safety data sheets are available for all reagents
Specifications
Photometry Tablet Reagents | |
Test kits, colorimeters and spectrophotometers | |
1 to 50mg/L | |
Molybdate No. 2 HR Tablet |