missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Tintometer™ Dissolved Oxygen Reagent No 3, 30mL
For use with Lovibond test kits, colorimeters and spectrophotometers
Brand: Lovibond™ 461170
Additional Details : Weight : 0.30000kg
Description
For use with Lovibond test kits, colorimeters and spectrophotometers
- Dissolved Oxygen Reagent No 3, 30mL
Specifications
Test kits, colorimeters and spectrophotometers | |
100 | |
30 mL | |
Dissolved Oxygen Reagent No 3 |