Learn More
Lovibond™ Tintometer™ CHECKIT™ Comparator, Copper Vario LR Test, 0 to 1mg/L
Suitable for both mobile and static water analysis. Checkit™ Comparators are available with a number of different color scales and provide the basis for a comprehensive easy to to to use analysis system.
Brand: Lovibond™ 147441
Additional Details : Weight : 0.74000kg
Description
- The Lovibond™ Checkit™ comparator is a compact, lightweight colorimetric unit
- The D55 Checkit Comparator contains mirror optics which extend the cell path length to 55mm, allowing greater accuracy at lower concentrations
- Suitable for both mobile and static water analysis
- Available with a number of different color scales
- Provides the basis for a comprehensive easy to to to use analysis system
- Each Checkit™ Comparator Disc contains a continuous color scale for exact color matching between standard and sample
- Checkit™ Comparator Copper LR Kit is supplied in a carry case with D55 Comparator, Vials, Discs, reagents and method instructions for all tests
- The Copper disc supplied measures from 0 to 1mg/L with the following steps: 0 / 0.05 / 0.1 / 0.15 / 0.2 / 0.25 / 0.3 / 0.35 / 0.4 / 0.45 / 0.5 / 0.6 / 0.7 / 0.8 / 0.9 / 1.0
Specifications
Water Quality Testing | |
Carry case with D55 Comparator, Vials, Discs, reagents and method instructions | |
Copper (free) Test |
Checkit™ Comparator D55 | |
0 to 10mg/L |