Learn More
Lovibond™ Tintometer™ Acidifying PT Rapid Tablets
For use with Lovibond test kits, colorimeters and spectrophotometers. Using tablet reagents guarantees exact reagent dosing, allowing for more precise titrations than traditional drop count test kits
382.85 NOK - 804.72 NOK
Specifications
For Use With (Application) | Photometry Tablet Reagents |
---|---|
For Use With (Equipment) | Test kits, colorimeters and spectrophotometers |
Quantity | 100 Tablets |
Type | Acidifying PT Rapid Tablets |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15311855
|
Lovibond™
515490BT |
100 Tablets |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15321855
|
Lovibond™
515491BT |
100 Tablets |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Acidifying PT Rapid Tablets
- Available in packs of 100 and 250
Specifications
Photometry Tablet Reagents | |
Test kits, colorimeters and spectrophotometers | |
100 Tablets | |
Acidifying PT Rapid Tablets |