missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TINAGL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12077-1-AP-150UL
Additional Details : Weight : 0.00010kg
Specifications
TINAGL1 | |
Polyclonal | |
Unconjugated | |
TINAGL1 | |
ARG1, GIS5, LCN7, LIECG3, OLRG 2, OLRG2, PSEC0088, TIN Ag related protein, TIN Ag RP, TINAGL, TINAGL1, TINAGRP, UNQ204/PRO230 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
64129, 94174, 94242 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q99JR5, Q9EQT5, Q9GZM7 | |
TINAGL1 | |
TINAGL1 Fusion Protein Ag2721 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |