missing translation for 'onlineSavingsMsg'
Learn More

TIGIT Rat anti-Mouse, eFluor® 660, Clone: GIGD7, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 50-9501-80

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15590097

  • 1124.00 NOK / 25µg


Estimated Shipment
08-09-2020
Add to basket

Description

Description

eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered. Binds with high affinity to the poliovirus receptor (PVR) which causes increased secretion of IL10 and decreased secretion of IL12B and suppresses T cell activation by promoting the generation of mature immunoregulatory dendritic cells.
Specifications

Specifications

TIGIT
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P86176
VSIG9, VSTM3, WUCAM, V-set and immunoglobulin domain containing 9, V-set and immunoglobulin domain-containing protein 9, V-set and transmembrane domain containing 3, V-set and transmembrane domain-containing protein 3, Washington University cell adhesion molecule
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
GIGD7
eFluor™ 660
Tigit
Liquid
Tigit
IgG2a, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
100043314
Videos
Documents

For Research Use Only.