missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thrombomodulin/BDCA-3 Antibody (501733) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3947JF669
4853.50 NOK valid until 2025-12-16
Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
Thrombomodulin/BDCA-3 Monoclonal antibody specifically detects Thrombomodulin/BDCA-3 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,ELISA Standard (Matched Pair),ELISA Capture (Matched Antibody Pair),ELISA Detection (Matched Antibody Pair),CyTOF
Specifications
| Thrombomodulin/BDCA-3 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| BDCA3, CD141, CD141 antigen, fetomodulin, thrombomodulin, TM | |
| Mouse myeloma cell line NS0-derived recombinant human Thrombomodulin/BDCA-3, Ala19-Ser515, Accession # P07204(Val473) | |
| 0.1 mL | |
| Cardiovascular Biology | |
| 7056 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, ELISA Standard (Matched Pair), ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair), CyTOF | |
| 501733 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction