Learn More
Thiosalicylic acid, 98%, Thermo Scientific Chemicals
Catalog Number p-7021173
Click to view available options
Quantity:
50 g
250 g
1000 g
147-93-3
C7H6O2S
154.183
MFCD00004836
NBOMNTLFRHMDEZ-UHFFFAOYSA-N
thiosalicylic acid, 2-mercaptobenzoic acid, o-mercaptobenzoic acid, 2-thiosalicylic acid, o-thiosalicylic acid, 2-carboxythiophenol, o-benzoic acid thiol, o-carboxythiophenol, o-sulfhydrylbenzoic acid, benzoic acid, 2-mercapto
5443
CHEBI:59124
2-sulfanylbenzoic acid
C1=CC=C(C(=C1)C(=O)O)S
147-93-3
C7H6O2S
154.183
MFCD00004836
NBOMNTLFRHMDEZ-UHFFFAOYSA-N
thiosalicylic acid, 2-mercaptobenzoic acid, o-mercaptobenzoic acid, 2-thiosalicylic acid, o-thiosalicylic acid, 2-carboxythiophenol, o-benzoic acid thiol, o-carboxythiophenol, o-sulfhydrylbenzoic acid, benzoic acid, 2-mercapto
5443
CHEBI:59124
2-sulfanylbenzoic acid
C1=CC=C(C(=C1)C(=O)O)S
Thiosalicylic acid acts as a trapping agent, which is useful in the desulfenylation of 3-indolyl sulfides. It is used to prepare macrocyclic diamides by condensation with diamines. Further, it is used as a precursor to drug, which finds application for the treatment of atherosclerosis and melanoma. In addition to this, it is involved in the preparation of vaccine preservative thiomersal and thioindigo dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Thiosalicylic acid acts as a trapping agent, which is useful in the desulfenylation of 3-indolyl sulfides. It is used to prepare macrocyclic diamides by condensation with diamines. Further, it is used as a precursor to drug, which finds application for the treatment of atherosclerosis and melanoma. In addition to this, it is involved in the preparation of vaccine preservative thiomersal and thioindigo dyestuff.
Solubility
Soluble in dimethyl sulfoxide, alcohol and ether. Slightly soluble in water, ethanol and alkanes.
Notes
Air and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 165°C to 168°C
Merck Index 149360
Solubility Information Soluble in dimethyl sulfoxide,alcohol and ether. Slightly soluble in water,ethanol and alkanes.
Formula Weight 154.19
Percent Purity 98%
Density 1.49
Quantity 50 g
Odor Stench
Sensitivity Air and light sensitive
Chemical Name or Material Thiosalicylic acid