missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiophene-3-carboxaldehyde diethyl acetal, 98%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Thiophene-3-carboxaldehyde diethyl aceta is used in OLED materials. Also used as electronic intermediate.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Thiophene-3-carboxaldehyde diethyl aceta is used in OLED materials. Also used as electronic intermediate.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Boiling Point | 130°C (30 mmHg) |
Refractive Index | 1.4865 |
Quantity | 1 g |
Beilstein | 125429 |
Solubility Information | Insoluble in water. |
Formula Weight | 186.27 |
Percent Purity | 98% |
Chemical Name or Material | Thiophene-3-carboxaldehyde diethyl acetal |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'