Learn More
Goat IgG Isotype Control, Invitrogen™
Goat Isotype Control
Brand: Invitrogen 31245
Additional Details : Weight : 0.01000kg
Description
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Purity/Antibody specificity: Based on immunoelectrophoresis at an antigen concentration of 20 mg/mL, the pattern of precipitation against rabbit anti-goat whole serum is the same as that against rabbit anti-goat IgG, Fc fragment specific. This antibody is recommended for use in blocking assays, immunohistochemical, immunocytochemical and immunofluorescent applications. Store product at 4°C until opened. After opening, dilute only enough product for a single day’s use. Store remainder at 4°C under sterile conditions. Country of Origin: USA
Goat IgG Control antibody specifically detects Goat IgG in Not Applicable samples. It is validated for Blocking Assay, Control, Immunocytochemistry, Immunohistochemistry.Specifications
Goat IgG | |
11.9 mg/mL | |
PBS with no preservative; pH 7.6 | |
Goat | |
10 mg | |
Isotype Control (Primary) | |
4° C | |
IgG |
Blocking Assay, Control, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry | |
Unconjugated | |
Immunoglobulin G; ImmunoglobulinG | |
Protein A | |
RUO | |
Not Applicable | |
Liquid |