Learn More
Thermo Scientific™ Forma™ Shelf Kit
Material: Copper; For Use With: Forma Steri-cycle CO2 Incubator, Forma Series 310 Direct heat CO2 Incubator, Forma series II Water jacketed CO2 Incubator
Brand: Thermo Scientific™ 190879
2343.76 NOK valid until 2024-03-29
Use promo code "21710" to get your promotional price.
Additional Details : Weight : 0.80000kg
Description
Precisely control the CO2 and temperature with Thermo Scientific™ Forma™ Series CO2 Incubators. Forma™ CO2 Incubator Accessories are important components for optimal operation.
- Precise control
- Choice of TC or IR sensors
- Superior parameter recovery characteristics
- Continuous contamination control technology
Specifications
TC Sensor or IR Sensor | |
LED | |
Shelf Kit |
Forma Steri-cycle CO2 incubator, Forma Series 310 Direct heat CO2 incubator, Forma series II Water jacketed CO2 incubator | |
Copper |