Learn More
Thermo Scientific™ Evolution™ UV-Visible Spectrophotometer to PC Interface Cable
Connects Evolution Series Spectrophotometers to laboratory PC
Brand: Thermo Scientific™ 401317263061
Additional Details : Weight : 1.00000kg
Includes: RS-232-C cable
Specifications
Interface Cable | |
Evolution™ series spectrophotometers | |
User manual and RS232C cable |