missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thalidomide, MP Biomedicals™
Click to view available options
Quantity:
100 mg
250 mg
50 g
Specifications
Color | White |
Quantity | 100 mg |
Physical Form | Solid |
Chemical Name or Material | Thalidomide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'