Learn More
Tetrahydropyran-2-methanol, 98%, Thermo Scientific Chemicals
Catalog Number p-3830475
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
100-72-1
C6H12O2
116.16
MFCD00006624
ROTONRWJLXYJBD-UHFFFAOYSA-N
tetrahydropyran-2-methanol, 2h-pyran-2-methanol, tetrahydro, 2-hydroxymethyltetrahydropyran, tetrahydropyran-2-carbinol, 2-hydroxymethyl tetrahydropyran, pyran-2-methanol, tetrahydro, tetrahydro-2h-pyran-2-yl methanol, 2-tetrahydropyranilcarbinol, tetrahydropyranyl-2-methanol, 2-methyloltetrahydro-1,4-pyran
7524
oxan-2-ylmethanol
C1CCOC(C1)CO
100-72-1
C6H12O2
116.16
MFCD00006624
ROTONRWJLXYJBD-UHFFFAOYSA-N
tetrahydropyran-2-methanol, 2h-pyran-2-methanol, tetrahydro, 2-hydroxymethyltetrahydropyran, tetrahydropyran-2-carbinol, 2-hydroxymethyl tetrahydropyran, pyran-2-methanol, tetrahydro, tetrahydro-2h-pyran-2-yl methanol, 2-tetrahydropyranilcarbinol, tetrahydropyranyl-2-methanol, 2-methyloltetrahydro-1,4-pyran
7524
oxan-2-ylmethanol
C1CCOC(C1)CO
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Density 1.0200g/mL
Boiling Point 187°C
Flash Point 93°C
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.4570 to 1.459
Quantity 10 g
Specific Gravity 1.02
Formula Weight 116.16
Percent Purity 98%
Chemical Name or Material Tetrahydropyran-2-methanol, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Tetrahydropyran-2-methanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.